Learn More
Crescent Chemical Co Inc EPA 515.4 METHYLDERV 16COMP1ML
SDP
Catalog No. 5068477
Encompass_Preferred
EPA Method 515.4 - Chlorinated Acids in Drinking Water by ECD, Quality Control Sample Methyl Derivatives, 16 components Acifluorfen methyl ester (50 ugmL), Bentazone methyl (100 ugmL), Chloramben methyl ester (50 ugmL), 2,4-Dimethyl ester (100ugmL),
Catalog No. 50-684-77 Supplier Crescent Chemical Co Inc Supplier No. CTCC515.4QC
EPA Method 515.4 - Chlorinated Acids in Drinking Water by ECD, Quality Control Sample Methyl Derivatives, 16 components Acifluorfen methyl ester (50 ugmL), Bentazone methyl (100 ugmL), Chloramben methyl ester (50 ugmL), 2,4-Dimethyl ester (100ugmL),