Learn More
Crescent Chemical Co Inc EPA 525.2 INTERN STD 3COMP 1ML
SDP
Catalog No. 5068374
Encompass_Preferred
EPA Method 525.2 - Internal Standard, 3 Components Acenaphthene d10, Chrysene d12, Phenanthrene d10 CAS Size - 1mL Catalog Number - CC2494
Catalog No. 50-683-74 Supplier Crescent Chemical Co Inc Supplier No. CTCC2494
EPA Method 525.2 - Internal Standard, 3 Components Acenaphthene d10, Chrysene d12, Phenanthrene d10 CAS Size - 1mL Catalog Number - CC2494