Learn More
Crescent Chemical Co Inc EPA 528 INTERN STD 2COMP 1ML
SDP
Catalog No. 5068489
Encompass_Preferred
EPA Method 528 Determination of Phenols in Drinking Water by Solid Phase Extraction Internal Std, varied conc in acetone, 2 comps-1,2-Dimethyl-3-nitrobenzene-250ug/mL,2,3,4,5-Tetrachlorophenol-500ug/mL 1mL
Catalog No. 50-684-89 Supplier Crescent Chemical Co Inc Supplier No. CTCC528IS
EPA Method 528 Determination of Phenols in Drinking Water by Solid Phase Extraction Internal Std, varied conc in acetone, 2 comps-1,2-Dimethyl-3-nitrobenzene-250ug/mL,2,3,4,5-Tetrachlorophenol-500ug/mL 1mL