Learn More
Crescent Chemical Co Inc EPA 531 .1MG/ML 11COMP 1ML
SDP
Catalog No. 5067894
Encompass_Preferred
EPA Method 531 - N-Methyl Carbamoyl Oximes and N-Methyl Carbamates, 11 components Aldicarb sulfoxide, Aldicarb sulfone, Oxamyl, Methomyl, 3-Hydroxycarbofuran, Aldicarb, Propoxur, Carbofuran, Carbaryl, Methiocarb, 1-Naphthol CAS Size
Catalog No. 50-678-94 Supplier Crescent Chemical Co Inc Supplier No. CTCC2080
EPA Method 531 - N-Methyl Carbamoyl Oximes and N-Methyl Carbamates, 11 components Aldicarb sulfoxide, Aldicarb sulfone, Oxamyl, Methomyl, 3-Hydroxycarbofuran, Aldicarb, Propoxur, Carbofuran, Carbaryl, Methiocarb, 1-Naphthol CAS Size