Learn More
Crescent Chemical Co Inc EPA 549.1 STDS 2COMP 1ML
SDP
Catalog No. 5068309
Encompass_Preferred
EPA Method 549.1 - Diquat and Paraquat Standard, 2 Components Diquat dibromide H20-1.97 mgmL, Paraquat dichloride 4H20-1.77 mgmL CAS Size - 1mL Catalog Number - CC2420A
Catalog No. 50-683-09 Supplier Crescent Chemical Co Inc Supplier No. CTCC2420A
EPA Method 549.1 - Diquat and Paraquat Standard, 2 Components Diquat dibromide H20-1.97 mgmL, Paraquat dichloride 4H20-1.77 mgmL CAS Size - 1mL Catalog Number - CC2420A