Learn More
Crescent Chemical Co Inc EPA 551.1 PEST/HERB 17COMP1ML
SDP
Catalog No. 5068504
Encompass
EPA Method 551.1 PesticidesHerbicide Mix, 17 Components Alachlor-10 ugmL, Atrazine-200 ugmL, Bromacil-10 ugmL, Cyanazine-30 ugmL, Endrin-2 ugmL, Endrin aldehyde-2 ugmL, Endrin ketone-2 ugmL, Heptachlor-1 ugmL, Heptachlor epoxide(B)-1 ugmL, Hexachloro
Catalog No. 50-685-04 Supplier Crescent Chemical Co Inc Supplier No. CTCC551.1PHM
EPA Method 551.1 PesticidesHerbicide Mix, 17 Components Alachlor-10 ugmL, Atrazine-200 ugmL, Bromacil-10 ugmL, Cyanazine-30 ugmL, Endrin-2 ugmL, Endrin aldehyde-2 ugmL, Endrin ketone-2 ugmL, Heptachlor-1 ugmL, Heptachlor epoxide(B)-1 ugmL, Hexachloro