Learn More
Crescent Chemical Co Inc EPA 554 CARBONYLS 12COMP 1ML
SDP
Catalog No. 5067947
Encompass_Preferred
EPA Method 554 - Carbonyl Compounds as DNPH Derivatives by HPLC, 12 Components Acetaldehyde, Butanal, Crotonaldehyde, Cyclohexanone, Decanal, Formaldehyde, Heptanal, Hexanal, Nonanal, Octanal, Pentanal, Propanal CAS Size - 1mL Cat
Catalog No. 50-679-47 Supplier Crescent Chemical Co Inc Supplier No. CTCC2111
EPA Method 554 - Carbonyl Compounds as DNPH Derivatives by HPLC, 12 Components Acetaldehyde, Butanal, Crotonaldehyde, Cyclohexanone, Decanal, Formaldehyde, Heptanal, Hexanal, Nonanal, Octanal, Pentanal, Propanal CAS Size - 1mL Cat