Learn More
Crescent Chemical Co Inc EPA 600 INTERNAL STD 6COMP 1ML
SDP
Catalog No. 5068306
Encompass_Preferred
CLP Semivolatile Priority Pollutant Standards - Internal Standards Mix, 6 Components Acenaphthene-d10, Chrysene-d12, 1,4-Dichlorobenzene-d4, Naphthalene-d8, Perylene-d12, Phenanthrene-d 10 CAS Size - 1mL Catalog Number - CC2418
Catalog No. 50-683-06 Supplier Crescent Chemical Co Inc Supplier No. CTCC2418
CLP Semivolatile Priority Pollutant Standards - Internal Standards Mix, 6 Components Acenaphthene-d10, Chrysene-d12, 1,4-Dichlorobenzene-d4, Naphthalene-d8, Perylene-d12, Phenanthrene-d 10 CAS Size - 1mL Catalog Number - CC2418