Learn More
Crescent Chemical Co Inc EPA 610 PAHS 16COMP 1ML
SDP
Catalog No. 5067953
Encompass_Preferred
EPA Method 8310 - Polynuclear Aromatic Hydrocarbons QC Mix, 16 Compounds Acenaphthene-0.1 mgmL, Acenaphthylene-0.1 mgmL, Anthracene-0.1 mgmL, Benz(a)anthracene-0.01 mgmL, Benzo(a)pyrene-0.01 mgmL, Benzo(b)fluoranthene-0.01 mgmL, Benzo(k)fluoranthene-
Catalog No. 50-679-53 Supplier Crescent Chemical Co Inc Supplier No. CTCC2113A
EPA Method 8310 - Polynuclear Aromatic Hydrocarbons QC Mix, 16 Compounds Acenaphthene-0.1 mgmL, Acenaphthylene-0.1 mgmL, Anthracene-0.1 mgmL, Benz(a)anthracene-0.01 mgmL, Benzo(a)pyrene-0.01 mgmL, Benzo(b)fluoranthene-0.01 mgmL, Benzo(k)fluoranthene-