Learn More
EPA 610 Polynuclear Aromatic Hydrocarbons Mix, Certified Reference Material, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| Synonym | EPA 610 Polynuclear Aromatic Hydrocarbons Mixture |
| Type | Food and Cosmetic Component Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | Acenaphthene 1000 μg/mL; Acenaphthylene 2000 μg/mL; Anthracene 100 μg/mL; Benz[a]anthracene 100 μg/mL; Benzo[b]fluoranthene 200 μg/mL; Benzo[k]fluoranthene 100 μg/mL; Benzo[ghi]perylene 200 μg/mL; Benzo[a]pyrene 100 μg/mL; Chrysene 100 μg/mL; Dibenz[a,h]anthracene 100 μg/mL; Fluoranthene 200 μg/mL; Fluorene 200 μg/mL;Indeno[1,2,3-cd]pyrene 100 μg/mL; Naphthalene 1000 μg/mL; Phenanthrene 100 μg/mL; Pyrene 100 μg/mL |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.