Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPA 625 Semivolatile Calibration Mix, 1,000 μg/mL, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111016410 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6410 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-6410 Supplier MilliporeSigma Supelco Supplier No. 506559
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy

1,000 μg/mL Each Component in Methylene chloride: Benzene (3:1)

TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Concentration 1000 μg/mL each component in methylene chloride: benzene (3:1)
Type Organic Pollutant Standard
Quantity 1 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) Acenaphthene; Acenaphthylene; Anthracene; Azobenzene; Benz[a]anthracene; Benzo[b]fluoranthene; Benzo[k]fluoranthene; Benzo[ghi]perylene; Benzo[a]pyrene; Benzyl butyl phthalate; Bis(2-chloroethoxy)methane; Bis(2-chloroethyl) ether; Bis(2-ethylhexyl) phthalate; 4-Bromodiphenyl ether; Carbazole; 4-Chlorodiphenyl ether; Bis-(2-chloroisopropyl) ether; 4-Chloro-3-methylphenol; 2-Chloronaphthalene; 2-Chlorophenol; Chrysene; Dibenz[a,h]anthracene; Dibutyl phthalate; 1,2-Dichlorobenzene; 1,3-Dichlorobenzene; 1,4-Dichlorobenzene; 2,4-Dichlorophenol; Diethyl phthalate; 2,4-Dimethylphenol; Dimethyl phthalate; 2,4-Dinitrophenol; 2,4-Dinitrotoluene; 2,6-Dinitrotoluene; Di-n-octyl phthalate; Fluoranthene; Fluorene; Hexachlorobenzene; Hexachloro-1,3-butadiene; Hexachlorocyclopentadiene; Hexachloroethane;Indeno[1,2,3-cd]pyrene; Isophorone; 2-Methyl-4,6-dinitrophenol; Naphthalene; Nitrobenzene; 2-Nitrophenol; 4-Nitrophenol; N-Nitrosodimethylamine; N-Nitrosodi-n-propylamine; Pentachlorophenol; Phenanthrene; Phenol; Pyrene; 1,2,4-Trichlorobenzene; 2,4,6-Trichlorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.