Learn More
Crescent Chemical Co Inc EPA 8081 DECOMP CHK 2COMP 1ML
SDP
Catalog No. 5072296
Encompass_Preferred
EPA Method 8081 - Decomposition Check Solution, 2 components 4,4-DDT, Endrin CAS Size - 1mL Catalog Number - 8DS-001
Catalog No. 50-722-96 Supplier Crescent Chemical Co Inc Supplier No. CT8DS001
EPA Method 8081 - Decomposition Check Solution, 2 components 4,4-DDT, Endrin CAS Size - 1mL Catalog Number - 8DS-001