Learn More
Crescent Chemical Co Inc EPA 8140 ORGANOPHOS 20COMP 1ML
SDP
Catalog No. 5068113
Encompass_Preferred
EPA Method 8140, Organophosphorous Pesticides, 20 Components Azinphos methyl, Bolstar, Chloropyrifos, Coumaphos, Demeton, Diazinon, Dichlorvos, Disulfoton, Ethoprop, Fensulfothion, Fenthion, Merphos, Methyl parathion, Mevinphos, Naled, Phorate, Ronne
Catalog No. 50-681-13 Supplier Crescent Chemical Co Inc Supplier No. CTCC2218
EPA Method 8140, Organophosphorous Pesticides, 20 Components Azinphos methyl, Bolstar, Chloropyrifos, Coumaphos, Demeton, Diazinon, Dichlorvos, Disulfoton, Ethoprop, Fensulfothion, Fenthion, Merphos, Methyl parathion, Mevinphos, Naled, Phorate, Ronne