Learn More
Crescent Chemical Co Inc EPA 8151 CHLOR ACIDS 18COMP1ML
SDP
Catalog No. 5068317
Encompass_Preferred
EPA Method 8151, Chlorinated Herbicides Acids, 18 Components Acifluorfen-0.1 mgmL, Bentazone-0.1 mgmL, Chloramben-0.1 mgmL, 2,4-D-0.1 mgmL, Dalapon-0.1 mgmL, 2,4-DB-0.1 mgmL, DCPA-0.1 mgmL, Dicamba-0.1 mgmL, 3,5-Dichlorobenzoic acid-0.1 mgmL, Dichlor
Catalog No. 50-683-17 Supplier Crescent Chemical Co Inc Supplier No. CTCC2440
EPA Method 8151, Chlorinated Herbicides Acids, 18 Components Acifluorfen-0.1 mgmL, Bentazone-0.1 mgmL, Chloramben-0.1 mgmL, 2,4-D-0.1 mgmL, Dalapon-0.1 mgmL, 2,4-DB-0.1 mgmL, DCPA-0.1 mgmL, Dicamba-0.1 mgmL, 3,5-Dichlorobenzoic acid-0.1 mgmL, Dichlor