Learn More
Crescent Chemical Co Inc EPA 8270 BASE/NEUT 6COMP 5ML
SDP
Catalog No. 5068194
Encompass_Preferred
EPA Method 8270, BaseNeutrals & Acids Matrix Spiking Solution, BaseNeutrals, 6 components Acenaphthene, 1,4-Dichlorobenzene, 2,4-Dinitrotoluene, N-Nitrosodi-n-propylamine, Pyrene, 1,2,4-Trichlorobenzene CAS Size - 5mL Catalog Numb
Catalog No. 50-681-94 Supplier Crescent Chemical Co Inc Supplier No. CTCC2291A5ML
EPA Method 8270, BaseNeutrals & Acids Matrix Spiking Solution, BaseNeutrals, 6 components Acenaphthene, 1,4-Dichlorobenzene, 2,4-Dinitrotoluene, N-Nitrosodi-n-propylamine, Pyrene, 1,2,4-Trichlorobenzene CAS Size - 5mL Catalog Numb