Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPA 8270 Calibration Mix 1, 1,000 μg/mL, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111016628 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6628 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6628 Supplier MilliporeSigma Supelco Supplier No. 5M00720
Add to Cart
Add to Cart

1,000 μg/mL Each Component in Dichloromethane : Benzene (3:1)

TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Concentration 1000 μg/mL each component in dichloromethane : benzene (3:1)
Type Organic Pollutant Standard
Quantity 5 x 1 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) Acenaphthene; Acenaphthylene; Anthracene; Azobenzene; Benz[a]anthracene; Benzo[b]fluoranthene; Benzo[k]fluoranthene; Benzo[ghi]perylene; Benzo[a]pyrene; Benzyl butyl phthalate; Bis(2-chloroethoxy)methane; Bis(2-chloroethyl) ether; Bis(2-ethylhexyl) phthalate; 4-Bromodiphenyl ether; Carbazole; 4-Chloroaniline; 4-Chlorodiphenyl ether; Bis-(2-chloroisopropyl) ether; 4-Chloro-3-methylphenol; 2-Chloronaphthalene; 2-Chlorophenol; Chrysene; p-Cresol; Dibenz[a,h]anthracene; Dibenzofuran; Dibutyl phthalate; 1,2-Dichlorobenzene; 1,3-Dichlorobenzene; 1,4-Dichlorobenzene; 2,4-Dichlorophenol; Diethyl phthalate; 2,4-Dimethylphenol; Dimethyl phthalate; 2,4-Dinitrophenol; 2,4-Dinitrotoluene; 2,6-Dinitrotoluene; Di-n-octyl phthalate; Fluoranthene; Fluorene; Hexachlorobenzene; Hexachloro-1,3-butadiene; Hexachlorocyclopentadiene; Hexachloroethane;Indeno[1,2,3-cd]pyrene; Isophorone; 2-Methyl-4,6-dinitrophenol; 2-Methylnaphthalene; o-Cresol; Naphthalene; 2-Nitroaniline; 3-Nitroaniline; 4-Nitroaniline; Nitrobenzene; 2-Nitrophenol; 4-Nitrophenol; N-Nitrosodimethylamine; N-Nitrosodi-n-propylamine; Pentachlorophenol; Phenanthrene; Phenol; Pyrene; 1,2,4-Trichlorobenzene; 2,4,5-Trichlorophenol; 2,4,6-Trichlorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.