Learn More
Crescent Chemical Co Inc EPA 8270 SV SURR STD 6COMP 1ML
SDP
Catalog No. 5068139
Encompass_Preferred
EPA Method 8270, Semivolatile Surrogate Standards Mix, 6 Components 2-Fluorobiphenyl, 2-Fluorophenol, Nitrobenzene d5, Phenol d6, 4-Terphenyl d14, 2,4,6-Tribromophenol CAS Size - 1mL Catalog Number - CC2258
Catalog No. 50-681-39 Supplier Crescent Chemical Co Inc Supplier No. CTCC2258
EPA Method 8270, Semivolatile Surrogate Standards Mix, 6 Components 2-Fluorobiphenyl, 2-Fluorophenol, Nitrobenzene d5, Phenol d6, 4-Terphenyl d14, 2,4,6-Tribromophenol CAS Size - 1mL Catalog Number - CC2258