Learn More
Crescent Chemical Co Inc EPA 8310 TARGET PAHS 16COMP1ML
SDP
Catalog No. 5068142
Encompass_Preferred
EPA Method 8310, Polynuclear Aromatic Hydrocarbons, Target PAH's, 16 Components Acenaphthene, Acenaphthylene, Anthracene, Benz(a)anthracene, Benzo(a)pyrene, Benzo(b)fluoranthene, Benzo(k)fluoranthene, Benzo(g,h,i)perylene, Chrysene, Dibenz(a,h)anthra
Catalog No. 50-681-42 Supplier Crescent Chemical Co Inc Supplier No. CTCC2264
EPA Method 8310, Polynuclear Aromatic Hydrocarbons, Target PAH's, 16 Components Acenaphthene, Acenaphthylene, Anthracene, Benz(a)anthracene, Benzo(a)pyrene, Benzo(b)fluoranthene, Benzo(k)fluoranthene, Benzo(g,h,i)perylene, Chrysene, Dibenz(a,h)anthra