Learn More
Cole-Parmer Cole-Parmer Precleaned EPA Amber Glass Wide-Mouth Bottle, 125 mL, 12/cs
Catalog No. NC1599144 Shop All Cole-Parmer Products
Encompass_Preferred
Glass bottles are processed and certified to meet the requirements of the current US EPA "Specifications and Guidance for Contaminant-Free Sample Containers" document for semi-volatile organics, pesticides, PCBs, metals, cyanide, fluoride, nitrate and nitrite analyses. Include PTFE-lined caps.
Catalog No. NC1599144 Supplier Cole-Parmer Supplier No. 9954051
Glass bottles are processed and certified to meet the requirements of the current US EPA "Specifications and Guidance for Contaminant-Free Sample Containers" document for semi-volatile organics, pesticides, PCBs, metals, cyanide, fluoride, nitrate and nitrite analyses. Include PTFE-lined caps.