Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPA method 1664 precision, accuracy and recovery standard, 4000μg/mL in acetone, Ultra Scientific

Catalog No. usrgo101x
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USRGO-101X 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USRGO-101X Supplier Agilent Technologies Supplier No. RGO101X
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material EPA method 1664 precision, accuracy and recovery standard
CAS Max % 98.99, 0.51, 0.51
CAS 544-76-3, 57-11-4, 67-64-1
Analyte or Component Names Acetone,n-hexadecane,octadecanoic acid (stearic acid)
Element Acetone, n-hexadecane, Octadecanoic Acid
Solvent or Matrix Acetone
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 4000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.