Learn More
Waters Corp Waters Corporation Analysis Kit, EPA Method 1694, Pharmaceuticals in Drinking Water
Catalog No. 50780054 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The EPA Method 1694 Analysis Kit streamlines the analysis of drinking water for pharmaceuticals. This kit includes the necessary components specified in EPA Method 1694 for the testing of Pharmaceuticals and Personal Care Products in Water, Soil, Sediment, and Biosolids by HPLC/MS/MS.
  • Designed for EPA methods applications.
  • Contains Oasis HLB cartridges, XTerrra HPLC columns, and Atlantis HPLC columns.
  • Simplifies the analysis of drinking water for pharmaceuticals.
  • Facilitates testing of pharmaceuticals and personal care products in water, soil, sediment, and biosolids.
Catalog No. 50-780-054 Supplier Waters Corp Supplier No. 176001634
The EPA Method 1694 Analysis Kit streamlines the analysis of drinking water for pharmaceuticals. This kit includes the necessary components specified in EPA Method 1694 for the testing of Pharmaceuticals and Personal Care Products in Water, Soil, Sediment, and Biosolids by HPLC/MS/MS.
  • Designed for EPA methods applications.
  • Contains Oasis HLB cartridges, XTerrra HPLC columns, and Atlantis HPLC columns.
  • Simplifies the analysis of drinking water for pharmaceuticals.
  • Facilitates testing of pharmaceuticals and personal care products in water, soil, sediment, and biosolids.