Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPA Method 200.7 Interference Check Standard No. 1, Ultra Scientific

Catalog No. USICM221
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-ICM-221 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-ICM-221 Supplier Agilent Technologies Supplier No. ICM221
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Chemical Name or Material EPA Method 200.7 Interference Check Standard No. 1
CAS 7697-37-2, 7732-18-5, 7439-98-7, 7440-21-3, 7440-32-6
CAS Max % 99.78, 0.01, 0.05, 0.03, 0.02, 0.1
Analyte or Component Names Water,Hydrofluoric acid,Boron
Element Hydrofluoric Acid, B
Solvent or Matrix Water/Trace Hydrofluoric Acid
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.