Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPA Method 200.7 Mixed Calibration Standard I, 10 Analytes, Ultra Scientific

Catalog No. USICM231
Encompass
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USICM231 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USICM231 Supplier Agilent Technologies Supplier No. ICM231
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Mixed Calibration Standard I
Name Note EPA Method 200.7
CAS Max % 97.96, 2.0, 0.0, 0.01, 0.0, 0.0
CAS 7440-36-0, 7440-38-2, 7440-39-3, 7697-37-2, 7732-18-5
Physical Form Liquid
Quantity 125 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.