Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPA Method 504.1 Mixture, 200μg/mL in methanol, Ultra Scientific

Catalog No. USDWM5141
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USDWM-514-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USDWM-514-1 Supplier Agilent Technologies Supplier No. DWM5141
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Chemical Name or Material EPA Method 504.1 Mixture
CAS Max % 99.92, 0.02, 0.02, 0.02
CAS 96-12-8, 106-93-4, 96-18-4
Analyte or Component Names methanol,1,2-dibromo-3-chloropropane (DBCP)
Element Methanol, 1,2-Dibromo-3-chloropropane
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 200μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.