Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPA PCB Congener Calibration Check Solution, 20 Analytes, Ultra Scientific

Catalog No. USRPCEPA1
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-RPC-EPA1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-RPC-EPA1 Supplier Agilent Technologies Supplier No. RPCEPA1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Chemical Name or Material EPA PCB Congener Calibration Check Solution
Name Note 0.2μg/mL
CAS 540-84-1, 34883-43-7, 37680-65-2, 7012-37-5, 41464-39-5
Physical Form Liquid
Color Colorless
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 0.2μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.