Learn More
Cole-Parmer EPA SAMPLE JARS, 60 ML, 24/CS
Supplier:  Cole-Parmer 9954040
Encompass_Preferred
Precleaned EPA Sample Jars, 60 mL, 24/csCertified clean for worry-free samplingInclude certificate of conformance to EPA Specifications and Guidance for Contaminant-Free Sample ContainersSample analysis labels included (unattached)PTFE-lined caps protect contents
Catalog No. NC1933274