Learn More
Phenomenex Inc EPA Water Analysis KG-33 Borosilicate Glass
Catalog No. 502394030 Shop All Phenomenex Products
Encompass_Preferred
EPA Water Analysis KG-33 Borosilicate Glass Vials w/o Closure 40mL Capacity 40mm L 24-400
Catalog No. 50-239-4030 Supplier Phenomenex Inc Supplier No. 3328952813
EPA Water Analysis KG-33 Borosilicate Glass Vials w/o Closure 40mL Capacity 40mm L 24-400