Learn More
Crescent Chemical Co Inc EPA600 BASE/NEU MIX1 14COMP1ML
SDP
Catalog No. 5068273
Encompass_Preferred
CLP Semivolatile Priority Pollutant Standards, BaseNeutrals Mix 1, 14 Components Bis(2-chloroethoxy)methane, Bis(2-chloroethyl)ether, Bis(2-chloroisopropyl)ether, Bis(2-ethylhexyl)phthalate, 4-Bromophenylphenyl ether, Butyl benzyl phthalate, 4-Chlor
Catalog No. 50-682-73 Supplier Crescent Chemical Co Inc Supplier No. CTCC2400
CLP Semivolatile Priority Pollutant Standards, BaseNeutrals Mix 1, 14 Components Bis(2-chloroethoxy)methane, Bis(2-chloroethyl)ether, Bis(2-chloroisopropyl)ether, Bis(2-ethylhexyl)phthalate, 4-Bromophenylphenyl ether, Butyl benzyl phthalate, 4-Chlor