Learn More
Crescent Chemical Co Inc EPA600 BASE/NEU MIX2 14COMP1ML
SDP
Catalog No. 5068278
Encompass_Preferred
CLP Semivolatile Priority Pollutant Standards, BaseNeutrals Mix 2, 14 Components Azobenzene, 2-Chloronaphthalene, 1,2-Dichlorobenzene, 1,3-Dichlorobenzene, 1,4-Dichlorobenzene, 2,4-Dinitrotoluene, 2,6-Dinitrotoluene, Hexachlorobenzene, Hexachlorobuta
Catalog No. 50-682-78 Supplier Crescent Chemical Co Inc Supplier No. CTCC2402
CLP Semivolatile Priority Pollutant Standards, BaseNeutrals Mix 2, 14 Components Azobenzene, 2-Chloronaphthalene, 1,2-Dichlorobenzene, 1,3-Dichlorobenzene, 1,4-Dichlorobenzene, 2,4-Dinitrotoluene, 2,6-Dinitrotoluene, Hexachlorobenzene, Hexachlorobuta