Learn More
Cayman Chemical Epacadostat, Purity: ≥98%, CAS: 1204669-58-8, 10 mg
SDP
Catalog No. 502981322
Encompass_Preferred
A selective IDO1 inhibitor (IC50 = 7.1 nM in HeLa cells); promotes T and natural killer-cell growth, increases IFN-γ production, and reduces conversion to Treg-like cells in a coculture system of human allogeneic lymphocytes with either dendritic cells or tumor cells; inhibits tumor growth in tumor-bearing mice in a lymphocyte-dependent manner
Catalog No. 50-298-1322 Supplier Cayman Chemical Supplier No. 1987510 MG
A selective IDO1 inhibitor (IC50 = 7.1 nM in HeLa cells); promotes T and natural killer-cell growth, increases IFN-γ production, and reduces conversion to Treg-like cells in a coculture system of human allogeneic lymphocytes with either dendritic cells or tumor cells; inhibits tumor growth in tumor-bearing mice in a lymphocyte-dependent manner