Learn More
Medchemexpress LLC HY-15689 5mg Medchemexpress, Epacadostat CAS:1204669-58-8 Purity:>98%
SDP
Catalog No. 502029158
Encompass_Preferred
Medchemexpress, HY-15689 5mg Epacadostat CAS:1204669-58-8 Epacadostat (INCB 024360) is a potent and selective indoleamine 2,3-dioxigenase 1 (IDO1) inhibitor with an IC50 of 71.8 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9158 Supplier Medchemexpress LLC Supplier No. HY156895MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-15689 5mg Epacadostat CAS:1204669-58-8 Epacadostat (INCB 024360) is a potent and selective indoleamine 2,3-dioxigenase 1 (IDO1) inhibitor with an IC50 of 71.8 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.