Learn More
Selleck Chemical LLC Epacadostat (INCB024360) 5mg 1204669-58-8
SDP
Supplier:  Selleck Chemical LLC S79105MG
Encompass_Preferred
Epacadostat (INCB024360) is a potent and selective indoleamine 2,3-dioxygenase (IDO1) inhibitor with IC50 of 10 nM and displays high selectivity over other related enzymes such as IDO2 or tryptophan 2,3-dioxygenase (TDO). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1200191