Learn More
Cayman Chemical EPA CYP450 Oxylipin MaxSpec LC-MS Mixture, 1 ea
SDP
Catalog No. 502995393
Encompass_Preferred
A mixture that contains (±)8(9)-EpETE, (±)11(12)-EpETE, (±)14(15)-EpETE, (±)17(18)-EpETE, (±)5(6)-DiHETE, (±)11(12)-DiHETE, (±)8(9)-DiHETE, (±)14(15)-DiHETE, and (±)17(18)-DiHETE
Catalog No. 50-299-5393 Supplier Cayman Chemical Supplier No. 213941 EA
A mixture that contains (±)8(9)-EpETE, (±)11(12)-EpETE, (±)14(15)-EpETE, (±)17(18)-EpETE, (±)5(6)-DiHETE, (±)11(12)-DiHETE, (±)8(9)-DiHETE, (±)14(15)-DiHETE, and (±)17(18)-DiHETE