Learn More
Apexbio Technology LLC Epalrestat 82159-09-9 200mg
SDP
Catalog No. 5000091763
Encompass_Preferred
Epalrestat (CAS 82159-09-9) is an orally available inhibitor of aldose reductase the rate-limiting enzyme of the polyol pathway involved in glucose metabolism By suppressing aldose reductase activity epalrestat limits intracellular sorbitol accumulation in neural tissues a process implicated in diabetic complications This compound is frequently employed in experimental models of diabetic neuropathy to investigate how modulation of the polyol pathway affects disease pathology and cellular signaling Epalrestat thus serves as a valuable molecular tool for elucidating the pathophysiology and pharmacology relevant to diabetes-associated neuronal damage
Catalog No. 50-000-91763 Supplier Apexbio Technology LLC Supplier No. B1743200
Epalrestat (CAS 82159-09-9) is an orally available inhibitor of aldose reductase the rate-limiting enzyme of the polyol pathway involved in glucose metabolism By suppressing aldose reductase activity epalrestat limits intracellular sorbitol accumulation in neural tissues a process implicated in diabetic complications This compound is frequently employed in experimental models of diabetic neuropathy to investigate how modulation of the polyol pathway affects disease pathology and cellular signaling Epalrestat thus serves as a valuable molecular tool for elucidating the pathophysiology and pharmacology relevant to diabetes-associated neuronal damage