Learn More
Sigma Aldrich Fine Chemicals Biosciences Epalrestat >=98% (HPLC) | 82159-09-9 | 10MG
Catalog No. 501868856
Encompass
Epalrestat >=98% (HPLC) | Purity: >=98% (HPLC) | Mol Wt: 319.4 | 82159-09-9 | 10MG
Catalog No. 50-186-8856 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SML052710MG
Epalrestat >=98% (HPLC) | Purity: >=98% (HPLC) | Mol Wt: 319.4 | 82159-09-9 | 10MG