Learn More
Adam Equipment EPB 363e - Equinox Precision Balances - Capacity: 360g, Readability: 0.001g
Catalog No. NC3964795
Encompass_Preferred
Equinox precision balances deliver remarkable precision to labs that perform highly detailed testing. The Equinox is ideal for research and quality assurance facilities; science education; precision counting; and production and manufacturing applications needing exceptionally precise results. Twelve precision models offer capacities from 360g to 8200g and readabilities of 1mg and 0.01g. A color touch screen display provides intuitive operation, with icon-driven menus and readily accessible applications and functions. The five-inch screen is generously sized, offering 800 X 480 pixel resolution for excellent visibility.
Catalog No. NC3964795 Supplier Adam Equipment Supplier No. EPB 363E
Equinox precision balances deliver remarkable precision to labs that perform highly detailed testing. The Equinox is ideal for research and quality assurance facilities; science education; precision counting; and production and manufacturing applications needing exceptionally precise results. Twelve precision models offer capacities from 360g to 8200g and readabilities of 1mg and 0.01g. A color touch screen display provides intuitive operation, with icon-driven menus and readily accessible applications and functions. The five-inch screen is generously sized, offering 800 X 480 pixel resolution for excellent visibility.