Learn More
Aviva Systems Biology Corp EPB41 Antibody - middle region (ARP42186_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42186_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EPB41. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EPB41. Alternate names: 4.1R, EL1, HEApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-58136