Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci EPB41L2 Peptide
SDP

Catalog No. 16823619
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-823-619 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-823-619 Supplier ProSci Supplier No. 45542P0100
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

EPB41L2 Peptide is used for blocking the activity of EPB41L2 antibody (catalog number 45-542). The EPB41L2 antibody ships at 0.5 mg/mL and the EPB41L2 Peptide ships as a 100 ug pellet. When peptide is reconstituted in 200 uL water, the concentration will be 0.5 mg/mL, identical to that of the antibody. The best starting ratio would be 1:1 (which means molar excess of peptide relative to antibody when identical volumes are mixed). Mix equal volume of peptide and antibody at the required dilution and leave at ambient temperature for 20 minutes. Then incubate and develop in parallel two blots: one with the antibody and one with the antibody which has been pre-absorbed with the provided peptide.

Specifications

Gene ID (Entrez) 2037
Species Human
Accession Number NP_001422.1
Common Name erythrocyte membrane protein band 4.1-like 2
Content And Storage Store at -20°C. When reconstituted minimize freezing and thawing.
Format Lyophilized
Formulation EPB41L2 peptide is supplied as 100 μg of dried peptide. When the peptide is reconstituted in 200 μL water, the concentration will be 0.5 mg/mL
For Use With (Application) Blocking Assay
Gene Symbol EPB41L2
Quantity 0.1 mg
Sequence EPB41L2 peptide corresponds to 11 amino acids of EPB41L2.
Synonym 4.1 G, 4.1-G
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.