Learn More
Aviva Systems Biology Corp EPC1 Antibody - middle region (ARP33343_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33343_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EPC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EPC1. Alternate names: DKFZp781P2312, Epl1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54970