Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4072MSM10P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: EGP40/1110. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: HT29 cells or Ovarian carcinoma. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: A recombinant human EpCAM fragment from the cytoplasmic domain. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 40-43kDa.Chromosome: 2p21. Gene Symbol: EPCAM. Gene ID: 4072. Ensemble: ENSG00000119888. Uniprot: NP_002345.
Catalog No. 50-188-4912