Learn More
Proteintech Group Inc EPCAM/CD326 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc 663161IG150UL
Encompass_Preferred
The EPCAM/CD326 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EPCAM/CD326. This antibody recognizes human antigen. The EPCAM/CD326 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Catalog No. 50-225-6507