Learn More
Enzo Life Sciences EpCAM (human) monoclonal antibody (VU-1D9) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804330C100
Encompass_Preferred
Clone: VU-1D9. Isotype: IgG1. Immunogen: Small cell lung carcinoma cells. Species reactivity: Human. Specificity: Recognizes human EpCAM (epithelial specific antigen), a 40 kDa epithelial protein expressed on baso-lateral cell surface in many epithelial tissues (but absent from mesothelial tissue). Differentiates between mesothelioma and carcinoma. Applications: Flow Cytometry, IHC (FS), IHC (PS), IP, WB, FUNC. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS.
Catalog No. 50-200-9867