Learn More
Sino Biological EpCAM Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90299C02H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus EPCAM [(Identical to the rhesus EPCAM (NP_001035118.1)] (Met1-Lys265) was expressed with the Fc region of human IgG1 at the C-terminus. Cynomolgus and Rhesus EPCAM sequences are identical.
Catalog No. 50-237-2799