Learn More
Sino Biological Human EpCAM / TROP-1 / TACSTD1 Protein (ECD, His Tag) 1mg
Supplier:  Sino Biological 10694H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Lys265) of human EpCAM (NP_002345.1) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-5484