Learn More
Sino Biological Human EpCAM / TROP-1 / TACSTD1 Protein (His Tag), Biotinylated 100ug
Supplier:  Sino Biological 10694H08HB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Lys265) of human EpCAM (NP_002345.1) was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6087