Learn More
Enzo Life Sciences EpCAM Recombinant monoclonal antibody (rMOC31) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS3170100
Encompass_Preferred
Clone: rMOC-31. Isotype: lgG1κ. Immunogen: Neuraminidase treated GLS-1 human small cell lung carcinoma cells. Species reactivity: Human. Specificity: Binding epitope of this antibody is located in the first EGF-like repeat domain (EGF1) between amino acids 27-59 of Ep-CAM. Applications: Flow Cytometry, IF, IHC (PS), WB. Purity: Protein G affinity purified. Formulation: Liquid. In 1X PBS containing 0.1 mg/ml BSA (US sourced) and 0.05% sodium azide.
Catalog No. 50-201-2848