Learn More
Enzo Life Sciences EpCAM Recombinant monoclonal antibody (EGP40/1556R) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS3650100
Encompass_Preferred
Clone: EGP40/1556R. Isotype: IgGκ. Immunogen: Recombinant human EpCAM. Species reactivity: Human. Applications: Flow Cytometry, IF, IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In 1X PBS containing 0.1 mg/ml BSA (US sourced) and 0.05% sodium azide.
Catalog No. 50-201-2893