Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate, Pentahydrate, Crystal, CP, Spectrum™ Chemical
SDP

Catalog No. 18610044 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-044 45 kg Poly Bottle
18-610-041 500 g Poly Bottle
18-610-042 2.5 kg Poly Bottle
18-610-043 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-610-044 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C143045KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7758-99-8 | 249.69 g/mol

Chemical Identifiers

CAS 7758-99-8

Specifications

Physical Form Crystalline Powder or Needles
CAS Max % 1
Quantity 45 kg
Packaging Poly Bottle
Molecular Weight (g/mol) 249.69
Grade Not Available
Chemical Name or Material Cupric Sulfate, Pentahydrate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.