Learn More
Sino Biological EpCAM / TROP-1 / TACSTD1 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 10694MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human EpCAM / TROP-1 / TACSTD1 (rh EpCAM / TROP-1 / TACSTD1; Catalog#10694-H08H; NP_002345.1; Met1-Lys265). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8127