Learn More
ACROBiosystems Recombinant Protein;Human EpCAM / TROP1 Protein, Fc Tag;HEK293;1MG;EPM,EPCAM,DIAR5,DIAR-5,EGP-2,EGP2,EGP314,EGP-314,EGP40,EGP-40,ESA,GA733-2,HNPCC8,HNPCC-8,KS1/4,KSA,M4S1,MIC18,MIC-18,MK-1,MK1,TACSTD1,TACSTD-1,TROP1,TROP-1,CD326,CD-326
Supplier:  ACROBiosystems EPMH52541MG
Encompass_Preferred
Recombinant Protein;Human EpCAM / TROP1 Protein, Fc Tag;1MG;Human EpCAM, Fc Tag (EPM-H5254) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 54.0 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7559